Learn More
Macherey-Nagel™ Chromabond™ HR-P Polypropylene SPE Column
Brand:  Macherey-Nagel™ 730108
 View more versions of this product
Product Code. 10503602
  • £96.40 / Pack of 30
Estimated Shipment: 21-11-2024
to see stock.
Explore more special offers
Add to basket
Specifications
3 mL
200 mg
Solid Phase Extraction Column
SPE
50 to 100 μm
1200 m2/g