Learn More
Macherey-Nagel™ Chromafix™ HR-XC Cartridge
Brand:  Macherey-Nagel™ 731756
 View more versions of this product
Product Code. 12801041
  • £219.00 / Pack of 50
Estimated Shipment: 23-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
Spherical PS/DVB Copolymer
45 or 85 μm
Chromafix HR-XC Cartridge
50 Pack
240 mg
Benzenesulfonic acid modified polymeric cation exchanger
65 to 75 Å
800 m2/g