missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Nichrome Inoculation Loops

Calibrated to remove a consistent, precise amount of material. Pro-Lab Diagnostics™ Nichrome Inoculation Loops are economical nickel-chrome wire loops in a range of sizes from 1μL to 10μL. The standard and heavy gauge wire loops can be used with insulated holders (sold separately).

Brand:  Pro-Lab Diagnostics™ PL.130

 View more versions of this product

Product Code. 12840155

  • £208.00 / Pack of 25
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

An inoculation loop is a simple tool used to gather and transfer microscopic organisms from a culture or broth plate. The small loop removes a consistent amount of inoculum by using surface tension. The inoculation loop is flame-sterilized before and after each use to eliminate cross-contamination between cultures. (The loop must be cooled after sterilization to avoid destroying the organisms.)

  • Constructed of economical nickel-chrome in standard or heavy gauge wire
  • Available in a range of 6 sizes, 1μL / 1.59mmD to 10μL / 5.05mmD
  • Packaged in sets of 25 loops
  • Holders with insulated handles sold separately
Specifications

Specifications

10 μL
Chrome-Nickel
Heavy
Yes
5.05 mm
Inoculating Loop
25/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pro-Lab Diagnostics™ Nichrome Inoculation Loops > Volume: 10uL; I.D.: 5.05mm; Heavy Gauge

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.