missing translation for 'onlineSavingsMsg'
Learn More
Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals Product Code.: 11356339

Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11356339
500 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11356339

Brand: Thermo Scientific Alfa Aesar 088059.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7789-02-8, 7697-37-2, 7732-18-5
Molecular Formula CrN3O9
Molecular Weight (g/mol) 238.01
MDL Number MFCD00010944
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
PubChem CID 9887206
IUPAC Name chromium(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

Especificaciones

Quantity 500 mL
Vapor Pressure 17mm Hg at 20°C
Physical Form Liquid
Color Colorless
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 5%; Chromium(III) nitrate nonahydrate: 0.77%; Water: 94.23%
Chemical Name or Material Chromium, AAS Standard Solution

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1521760 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog Number088059.AP,088059.AE
Lot Number1521761 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog Number088059.AP,088059.AE
Lot Number1510736 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog Number088059.AP,088059.AE
Lot Number1510737 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog Number088059.AP,088059.AE
Lot Number1496652 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number088059.AP,088059.AE

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado