missing translation for 'onlineSavingsMsg'
Learn More
Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals Product Code.: 11389149

Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11389149
100 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11389149

Brand: Thermo Scientific Alfa Aesar 088059.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-02-8, 7697-37-2, 7732-18-5
Molecular Formula CrN3O9
Molecular Weight (g/mol) 238.01
MDL Number MFCD00010944
InChI Key PHFQLYPOURZARY-UHFFFAOYSA-N
PubChem CID 9887206
IUPAC Name chromium(3+);trinitrate;nonahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3]

Specifications

Quantity 100 mL
Vapor Pressure 17mm Hg at 20°C
Physical Form Liquid
Color Colorless
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 5%; Chromium(III) nitrate nonahydrate: 0.77%; Water: 94.23%
Chemical Name or Material Chromium, AAS Standard Solution

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1521760 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog Number088059.AP,088059.AE
Lot Number1521761 Certificate TypeCertificate of Analysis Date14/01/2025 Catalog Number088059.AP,088059.AE
Lot Number1510736 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog Number088059.AP,088059.AE
Lot Number1510737 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog Number088059.AP,088059.AE
Lot Number1496652 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number088059.AP,088059.AE
compliance-icons
Product Identifier
  • Chromium, AAS standard solution, Specpure, Cr 1000
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium, AAS standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.