Learn More
Chromium(III) chloride, 99%, anhydrous, Thermo Scientific Chemicals
£15.58 - £134.67
Chemical Identifiers
CAS | 10025-73-7 |
---|---|
MDL Number | MFCD00010948 |
Synonym | chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0fShow More |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10185583
|
Thermo Scientific Acros
190470050 |
5 g | Glass bottle |
£15.58
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10793381
|
Thermo Scientific Acros
190470250 |
25 g | Plastic bottle |
£49.55
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10752564
|
Thermo Scientific Acros
190471000 |
100 g | Plastic bottle |
£134.67
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-73-7 | |
MFCD00010948 | |
chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f |
Specifications
1152.0°C | |
Purple | |
98.5% min | |
CrCl3 | |
15,95 | |
chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f | |
158.36 | |
99% | |
Chromium(III) chloride, 99% |
10025-73-7 | |
Shiny Fluffy Powder or Flakes | |
Cl3Cr | |
MFCD00010948 | |
15, 2226 | |
Solubility in water: slightly soluble | |
158.36 | |
2.76 |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
May be corrosive to metals.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Absorb spillage to prevent material damage.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SK
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 233-038-3
missing translation for 'rtecsNumber' : GB5425000
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.