missing translation for 'onlineSavingsMsg'
Learn More
Chromium(III) chloride, 99%, anhydrous, Thermo Scientific Chemicals Product Code.: 10793381

Chromium(III) chloride, 99%, anhydrous, Thermo Scientific Chemicals

Product Code. 10793381
25 g, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10793381

Brand: Thermo Scientific Acros 190470250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-73-7
MDL Number MFCD00010948
InChI Key LJAOOBNHPFKCDR-UHFFFAOYSA-K
Synonym chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0fShow MoreShow Less
IUPAC Name chromium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3]

Specifications

Melting Point 1152.0°C
Color Purple
Physical Form Shiny Fluffy Powder or Flakes
Quantity 25 g
Assay Percent Range 98.5% min
Molecular Formula Cl3CrH12O6
Linear Formula CrCl3
Fieser 15,95
Merck Index 15, 2226
Solubility Information Solubility in water: slightly soluble
Molecular Weight (g/mol) 266.44
Formula Weight 158.36
Percent Purity 99%
Packaging Plastic bottle
Density 2.76
Chemical Name or Material Chromium(III) chloride, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467300 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number190470050,190470250,190471000
Lot NumberA0466435 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number190470050,190470250,190471000
Lot NumberA0465403 Certificate TypeCertificate of Analysis Date19/08/2024 Catalog Number190470050,190470250,190471000
Lot NumberA0462835 Certificate TypeCertificate of Analysis Date27/05/2024 Catalog Number190470050,190470250,190471000
Lot NumberA0459439 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog Number190470050,190470250,190471000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium(III) chloride, 99%, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.