missing translation for 'onlineSavingsMsg'
Learn More
Chromium(III) potassium sulfate dodecahydrate, 98%, Thermo Scientific Chemicals Product Code.: 11354559

Chromium(III) potassium sulfate dodecahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11354559
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11354559

Brand: Thermo Scientific Alfa Aesar A11816.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chromium(III) potassium sulfate is used for tanning of leather and as a photographic film hardener. It is used in manufacturing of chromium-containing catalyst.It is used in mordantin in printing and dyeing industry and it could also be used as analytical reagent. In addition, it is also used as food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromium(III) potassium sulfate is used for tanning of leather and as a photographic film hardener. It is used in manufacturing of chromium-containing catalyst.It is used in mordantin in printing and dyeing industry and it could also be used as analytical reagent. In addition, it is also used as food additive.

Solubility
Soluble in water. 24 g/100 mL (15°C)

Notes
Incompatible with magnesium, aluminum. Store away from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7788-99-0
Molecular Formula CrH24KO20S2
Molecular Weight (g/mol) 499.386
MDL Number MFCD00149917
InChI Key ZFVHBEKVAITXHW-UHFFFAOYSA-J
Synonym chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4pShow MoreShow Less
PubChem CID 24596
IUPAC Name potassium;chromium(3+);disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]

Specifications

Melting Point 89°C
Quantity 100 g
Linear Formula CrK(SO4)2·12H2O
Merck Index 14,2229
Solubility Information Soluble in water. 24g/100 mL (15°C)
Formula Weight 499.39 (283.21 Anhydrous)
Percent Purity 98%
Odor Odorless
Density 1.83
Chemical Name or Material Chromium (III) potassium sulfate dodecahydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243046 Certificate TypeCertificate of Analysis Date08/09/2022 Catalog NumberA11816.0E,A11816.36,A11816.22
Lot Number10203993 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11816.0E,A11816.36,A11816.22
Lot Number10159333 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11816.0E,A11816.36,A11816.22
Lot Number10182908 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11816.0E,A11816.36,A11816.22
Lot NumberF3540A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11816.0E,A11816.36,A11816.22
compliance-icons
Product Identifier
  • Chromium(III) potassium sulfate dodecahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Chromium(III) potassium sulfate dodecahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium(III) potassium sulfate dodecahydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.