Learn More
Chromium(III) nitrate nonahydrate, 99%, Thermo Scientific Chemicals
£10.50 - £302.51
Chemical Identifiers
CAS | 7789-02-8 |
---|---|
MDL Number | MFCD00149659 |
InChI Key | PHFQLYPOURZARY-UHFFFAOYSA-N |
Synonym | chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate |
PubChem CID | 9887206 |
IUPAC Name | chromium(3+);trinitrate;nonahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10511281
|
Thermo Scientific
219200050 |
5 g | Glass bottle | N/A | ||||||
10092830
|
Thermo Scientific Acros
219201000 |
100 g | Plastic bottle |
£18.29
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10124990
|
Thermo Scientific Acros
219205000 |
500 g | Plastic bottle |
£38.89
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10092820
|
Thermo Scientific Acros
219200010 |
1 kg | Plastic bottle |
£67.64
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10399123
|
Thermo Scientific Acros
219200051 |
5 kg | Plastic bottle |
£302.51
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-02-8 | |
PHFQLYPOURZARY-UHFFFAOYSA-N | |
9887206 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3] |
MFCD00149659 | |
chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate | |
chromium(3+);trinitrate;nonahydrate |
Specifications
60.0°C | |
Black to Purple | |
Crystalline Chunks | |
CrN3O9 | |
MFCD00149659 | |
Solubility in water: 1g/L (20°C). Other solubilities: soluble in alcohol | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Cr+3] | |
238.01 | |
400.14 | |
Chromium(III) nitrate nonahydrate, 99% |
7789-02-8 | |
2 to 3 (5% aq. soln.) | |
99% | |
Cr(NO3)3·9H2O | |
chromium iii nitrate nonahydrate, unii-d2806iol1l, nitric acid, chromium 3+ salt, nonahydrate, acmc-20alg5, bis 7-chlorophenyl sulfone, cr.3no3.9h2o, ksc495a9j, chromium trinitrate, nonahydrate, chromium iii nitrate, nonahydrate, chromium 3+ nonahydrate trinitronate | |
PHFQLYPOURZARY-UHFFFAOYSA-N | |
chromium(3+);trinitrate;nonahydrate | |
9887206 | |
99% |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes serious eye irritation.
May intensify fire; oxidizer.
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Take any precaution to avoid mixing with combustibles.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Danger
missing translation for 'rtecsNumber' : GB6300000
- Chromium(III) nitrate nonahydrate
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.