missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) phosphate tetrahydrate, Thermo Scientific Chemicals

For Use As a Catalyst, Pigment

£45.33 - £180.90

Chemical Identifiers

CAS 10101-59-4
Molecular Formula CrH8O8P
Molecular Weight (g/mol) 219.026
MDL Number MFCD00016049
InChI Key BVZHAJJRIRDSTG-UHFFFAOYSA-K
Synonym chromium iii phosphate tetrahydrate, cr.po4.4h2o, chromium 3+ ion tetrahydrate phosphate, chromium 3+ phosphate-water 1/1/4, chromium 3+ tetrahydrate phosphateShow MoreShow Less
PubChem CID 53471880
IUPAC Name chromium(3+);phosphate;tetrahydrate
SMILES O.O.O.O.[O-]P(=O)([O-])[O-].[Cr+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11372648
SDS View Documents
Thermo Scientific Alfa Aesar
024110.09
10 g
£45.33
10g
Please to purchase this item. Need a web account? Register with us today!
11382648
SDS View Documents
Thermo Scientific Alfa Aesar
024110.18
50 g
£180.90
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as an additive in drugs, anti-corrosive pigments, catalysts and polymer manufacture, is also used to catalyze cation exchange in sorption reactions, as a laminated structure to dampen vibration and noise in a motor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 232-141-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium(III) phosphate tetrahydrate, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.