missing translation for 'onlineSavingsMsg'
Learn More

Chromium, plasma standard solution, Specpure™ Cr 10,000 ^mg/ml, Thermo Scientific Chemicals

£68.54 - £262.31

Chemical Identifiers

MDL Number MFCD00010944
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11329007
View Documents
Thermo Scientific Alfa Aesar
038722.AD
50 mL
£68.54
50mL
Please to purchase this item. Need a web account? Register with us today!
11319007
View Documents
Thermo Scientific Alfa Aesar
038722.AE
100 mL
£104.52
100mL
Please to purchase this item. Need a web account? Register with us today!
11460171
View Documents
Thermo Scientific Alfa Aesar
038722.AP
500 mL
£262.31
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00010944

Specifications

Cr(NO3)3 in 5% HNO3
UN3264
Liquid
MFCD00010944
Specpure™
Chromium plasma standard solution, Cr 10000 μg/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P234-P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P390-P501c

H290-H314-H317-H335

missing translation for 'dotInformation' : Hazard Class: 8

missing translation for 'einecsNumber' : 231-714-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.