missing translation for 'onlineSavingsMsg'
Learn More

Chromium selenide, Black powder, 99.5% (Metals basis), Thermo Scientific Chemicals

£296.48

Chemical Identifiers

CAS 12053-13-3
Molecular Formula CrSe
Molecular Weight (g/mol) 130.967
MDL Number MFCD00049885
InChI Key UVZCKRKEVWSRGT-UHFFFAOYSA-N
Synonym chromium selenide, chromium ii selenide
PubChem CID 82886
IUPAC Name selanylidenechromium
SMILES [Cr]=[Se]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11392208
View Documents
Thermo Scientific Alfa Aesar
013119.14
25 g
£296.48
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used in semiconductor, chemical vapor deposition (CVD) and physical vapor deposition (PVD) display and optical applications. It is used for coating solar energy or fuel cells and flip-chip applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12053-13-3
130.967
UVZCKRKEVWSRGT-UHFFFAOYSA-N
82886
[Cr]=[Se]
CrSe
MFCD00049885
chromium selenide, chromium ii selenide
selanylidenechromium

Specifications

12053-13-3
Powder
CrSe
chromium selenide, chromium ii selenide
[Cr]=[Se]
130.967
130.96
6.74g/cm3
Black
99.5% (Metals basis)
MFCD00049885
UVZCKRKEVWSRGT-UHFFFAOYSA-N
selanylidenechromium
82886
Odorless
Chromium selenide
Videos
Safety and Handling

Safety and Handling

When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.

H301-H331-H373-H400-H410; Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

missing translation for 'dotInformation' : DOT Class: 6.1, Packaging Group: II

missing translation for 'einecsNumber' : 234-999-1

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only