Learn More
Chromium(VI) oxide, ACS, 98+%, Thermo Scientific Chemicals
£36.20 - £103.00
Chemical Identifiers
CAS | 1333-82-0 |
---|---|
Molecular Formula | CrO3 |
Molecular Weight (g/mol) | 99.99 |
MDL Number | MFCD00010952 |
InChI Key | WGLPBDUCMAPZCE-UHFFFAOYSA-N |
Synonym | chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide |
PubChem CID | 14915 |
ChEBI | CHEBI:48240 |
IUPAC Name | trioxochromium |
SMILES | O=[Cr](=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11347928
|
Thermo Scientific Alfa Aesar
036468.22 |
100 g |
£36.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11337928
|
Thermo Scientific Alfa Aesar
036468.36 |
500 g |
£103.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chromium plating, copper stripping, aluminum anodizing, corrosion inhibitor, photography, ceramic glazes, colored glass, oxidizing agent in organic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Chromium plating, copper stripping, aluminum anodizing, corrosion inhibitor, photography, ceramic glazes, colored glass, oxidizing agent in organic chemistry
Chemical Identifiers
1333-82-0 | |
99.99 | |
WGLPBDUCMAPZCE-UHFFFAOYSA-N | |
14915 | |
trioxochromium |
CrO3 | |
MFCD00010952 | |
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide | |
CHEBI:48240 | |
O=[Cr](=O)=O |
Specifications
250°C (decomposition) | |
1333-82-0 | |
>98+% | |
MFCD00010952 | |
14,2235 | |
Soluble in water,H2SO4,and alcohol; Also soluble in ether and nitric acid. | |
O=[Cr](=O)=O | |
99.99 | |
CHEBI:48240 | |
≥98% | |
Hygroscopic | |
Chromium oxide, For analysis ACS |
196°C | |
Crystalline | |
CrO3 | |
UN1463 | |
chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide | |
WGLPBDUCMAPZCE-UHFFFAOYSA-N | |
trioxochromium | |
14915 | |
99.99 | |
ACS | |
2.7 g/mL |
Safety and Handling
GHS H Statement
H271-H301-H310-H330-H334-H340-H350-H372-H361-H314-H318-H317
P201-P202-P210-P220-P221-P260-P262-P264b-P270-P271-P272-P280g-P281-P283-P284-P285-P302+P350-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P333+P313-P353-P363-P370+P378q-P3
H271-H301-H310+H330-H314-H317-H334-H335-H340-H350-H361f-H372
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: CHROMIUM TRIOXIDE, ANHYDROUS
missing translation for 'einecsNumber' : 215-607-8
missing translation for 'rtecsNumber' : GB6650000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only