missing translation for 'onlineSavingsMsg'
Learn More
Chromium(VI) oxide, ACS, 98+%, Thermo Scientific Chemicals Product Code.: 11337928

Chromium(VI) oxide, ACS, 98+%, Thermo Scientific Chemicals

Product Code. 11337928
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11337928

Brand: Thermo Scientific Alfa Aesar 036468.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Chromium plating, copper stripping, aluminum anodizing, corrosion inhibitor, photography, ceramic glazes, colored glass, oxidizing agent in organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Chromium plating, copper stripping, aluminum anodizing, corrosion inhibitor, photography, ceramic glazes, colored glass, oxidizing agent in organic chemistry
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxideShow MoreShow Less
PubChem CID 14915
ChEBI CHEBI:48240
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Boiling Point 250°C (decomposition)
Melting Point 196°C
Color Red
Physical Form Crystalline
Quantity 500 g
Assay Percent Range >98+%
UN Number UN1463
Merck Index 14,2235
Solubility Information Soluble in water,H2SO4,and alcohol; Also soluble in ether and nitric acid.
Formula Weight 99.99
Percent Purity ≥98%
Grade ACS
Sensitivity Hygroscopic
Density 2.7 g/mL
Chemical Name or Material Chromium(VI) oxide, For analysis ACS
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS07K040 Certificate TypeCertificate of Analysis Date17/06/2024 Catalog Number036468.22,036468.36
Lot NumberP22J004 Certificate TypeCertificate of Analysis Date12/12/2023 Catalog Number036468.22,036468.36
Lot NumberW29J014 Certificate TypeCertificate of Analysis Date12/12/2023 Catalog Number036468.22,036468.36
Lot NumberZ14I033 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog Number036468.22,036468.36
Lot NumberQ01I027 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number036468.22,036468.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium(VI) oxide, ACS, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.