missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromofil™ PTFE Syringe Filter

Filter nonpolar liquids and gases. Chromofil™ PTFE Syringe Filters with polytetrafluorethylne membrane come color coded.

Brand:  Macherey-Nagel™ 729007

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10496471

  • £111.00 / Pack of 100
Estimated Shipment: 20-08-2024
to see stock.
Add to basket

Description

Description

  • Syringe filters with polytetrafluoroethylene membrane  
  • Color-coded  
  • Hydrophobic membrane
  • For nonpolar liquids and gases
  • Very resistant towards all kinds of solvents as well as acids and bases
  • Flushing with alcohol, followed by water, makes the originally hydrophobic membrane more hydrophilic.
Specifications

Specifications

Yellow Top, Colorless Bottom
0.2 μm
Hydrophobic
Syringe Filter
25 mm
PTFE
Non-Polar Liquids and Gases
100 Pack
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers