Learn More
Proteintech ChromoTek Binding Control Magnetic Agarose
Binding Controls are deactivated magnetic agarose beads. They are the same magnetic agarose matrix as our Nano-Traps Magnetic Agarose.
Brand: Proteintech BMAB-20
Additional Details : Weight : 0.00240kg
Description
- Control of unspecific binding of proteins, DNA, etc. to beads
- Pre-clearing of cell lysate
- 20 reactions / 500 μL slurry
- Stored in 20% EtOH
Specifications
20 Reactions |