missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Myc-Trap™ Magnetic Agarose
The ChromoTek Myc-Trap™ Magnetic Agarose is an affinity resin for IP of Myc-fusion proteins.
£208.00 - £5160.00
Specifications
Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. |
---|---|
For Use With (Application) | IP, CoIP, ChIP, RIP |
Target | Myc-tag sequence motif EQKLISEEDL at the N-terminus, C-terminus, or internal site of the fusion protein. Endogenous c-myc is NOT bound. |
Label or Dye | Magnetic Agarose |
Species | Alpaca |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
17303583
|
Proteintech
YTMA-100 |
100 Reactions |
£1572.00
100 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17313583
|
Proteintech
YTMA-20 |
20 Reactions |
£348.00
20 reactions |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
17323583
|
Proteintech
YTMA-200 |
200 Reactions |
£2828.00
200 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17333583
|
Proteintech
YTMA-400 |
400 Reactions |
£5160.00
400 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17393573
|
Proteintech
YTMA-10 |
10 Reactions |
£208.00
10 reactions |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
Description
- Consists of a Myc Nanobody/VHH coupled to magnetic agarose beads
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. | |
Myc-tag sequence motif EQKLISEEDL at the N-terminus, C-terminus, or internal site of the fusion protein. Endogenous c-myc is NOT bound. | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose |