missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Spin Columns
Filtration units from PP and 20 μm PE frit for batch mode centrifugation based separation.
£26.00 - £93.00
Specifications
For Use With (Application) | Rapid washing and clean elution of bound proteins |
---|---|
Type | Spin Column |
Sample Volume | 500 μL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17303463
|
Proteintech
SCT-10 |
10/Pk. |
£26.00
Pack of 10 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
|||||
17313463
|
Proteintech
SCT-20 |
20/Pk. |
£41.00
Pack of 20 |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
17323463
|
Proteintech
SCT-50 |
50/Pk. |
£93.00
Pack of 50 |
Estimated Shipment: 04-07-2024
Log in to see stock. |
|||||
Description
- The spin columns allow rapid washing and clean elution of bound proteins.
Spin columns simplify the pull-down of your protein with Nano-Traps coupled to agarose beads - The spin columns are designed for easy and convenient handling: For washing, you may discard the flow-through wash buffer; for elution, collect the flow-through after adding the elution buffer
Specifications
Rapid washing and clean elution of bound proteins | |
Spin Column | |
500 μL |