missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek TurboGFP-Trap Magnetic Agarose
TurboGFP-Trap Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of TurboGFP fusion proteins.
£208.00 - £5160.00
Specifications
Content And Storage | Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. |
---|---|
For Use With (Application) | IP, CoIP, ELISA |
Target | TurboGFP, CopGFP, Pontellina plumata, maxGFP, maxFP-Green |
Label or Dye | Magnetic Agarose |
Species | Alpaca |
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sufficient For | Price | Quantity & Availability | |||||
17303523
|
Proteintech
TBTMA-10 |
10 Reactions |
£208.00
10 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17313523
|
Proteintech
TBTMA-100 |
100 Reactions |
£1572.00
100 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17323523
|
Proteintech
TBTMA-20 |
20 Reactions |
£348.00
20 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17333523
|
Proteintech
TBTMA-200 |
200 Reactions |
£2828.00
200 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
17343523
|
Proteintech
TBTMA-400 |
400 Reactions |
£5160.00
400 reactions |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
Description
- Comprises an anti-TurboGFP VHH/ Nanobody coupled to magnetic agarose beads
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. | |
TurboGFP, CopGFP, Pontellina plumata, maxGFP, maxFP-Green | |
Alpaca |
IP, CoIP, ELISA | |
Magnetic Agarose |