Learn More
Proteintech ChromoTek V5-Trap™ Magnetic Agarose
The ChromoTek V5-Trap™ Magnetic Agarose is an affinity resin for IP of V5-tagged proteins.
Brand: Proteintech V5TMA-200
Additional Details : Weight : 0.00240kg
Description
- Consists of a anti-V5 Nanobody/VHH coupled to magnetic agarose beads
Specifications
Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year. | |
V5-tag sequence GKPIPNPLLGLDST at the N-, C-terminal or internal tagging. | |
Alpaca |
IP, CoIP, ChIP, RIP | |
Magnetic Agarose | |
200 Reactions |