missing translation for 'onlineSavingsMsg'
Learn More

Chromplete™ 2-Propanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™

The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Isopropanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.

£106.00 - £256.00

Chemical Identifiers

CAS 67-63-0
Molecular Formula C3H8O
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
PubChem CID 3776
ChEBI CHEBI:17824
IUPAC Name propan-2-ol
SMILES CC(C)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15950721
View Documents Promotion Details
Thermo Scientific
T001041000
1 L
£162.00
1 L
Please to purchase this item. Need a web account? Register with us today!
15960721
View Documents
Thermo Scientific
T001042500
2.5 L
£256.00
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15970721
View Documents
Thermo Scientific
T001044000
4 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Chromplete Isopropanol meets the analytical requirements of USP and is suitable for use in:

  • HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
  • GC- FID, GC- ECD, GC-TCD
  • Pesticide Analysis by GC or HPLC
  • UV Spectrophotometry
  • Preparative chromatography
  • Supercritical Fluid and Thin Layer chromatography
  • Reactions/Analyses requiring ACS compliance
  • Extraction/Concentration Analyses
Specifications

Chemical Identifiers

67-63-0
KFZMGEQAYNKOFK-UHFFFAOYSA-N
CHEBI:17824
CC(C)O
C3H8O
3776
propan-2-ol

Specifications

-90°C
67-63-0
≤ 5 APHA
Amber Glass
≤ 1.0 ppm
1219
isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
CC(C)O
60.1
CHEBI:17824
99.9%
Minimum, GC-FID
≤ 1.0, ≤ 0.20, ≤ 0.10, ≤ 0.015
0.999
83°C
Liquid
C3H8O
≤0.05%
KFZMGEQAYNKOFK-UHFFFAOYSA-N
propan-2-ol
3776
60.1
Chromplete
2-Propanol
Videos
Safety and Handling

Safety and Handling

Causes serious eye irritation, Highly flammable liquid and vapor, May cause drowsiness or dizziness

Serious eye damage/eye irritation (category 2), Flammable liquids (category 2), Specific target organ toxicity after single exposure (category 3)

missing translation for 'shelfLife' : 1825 days

missing translation for 'einecsNumber' : 200-661-7

missing translation for 'rtecsNumber' : NT8050000

missing translation for 'storageNote' : At room temperature in sealed containers

Recommended Storage : Room temperature flammable storage

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only