Learn More
Chromplete™ Acetonitrile, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™
The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Acetonitrile is suitable for HPLC, GC, SFC, TLC, ACS, Spectrophotometry, and meets the analytical requirements of USP.
£59.95 - £258.00
Chemical Identifiers
CAS | 75-05-8 |
---|---|
Molecular Formula | C2H3N |
Molecular Weight (g/mol) | 41.05 |
InChI Key | WEVYAHXRMPXWCK-UHFFFAOYSA-N |
Synonym | methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn |
PubChem CID | 6342 |
ChEBI | CHEBI:38472 |
IUPAC Name | acetonitrile |
SMILES | CC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15945310
|
Thermo Scientific
T001011000 |
1 L | Aluminum Jug |
£258.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955310
|
Thermo Scientific
T001012500 |
2.5 L | Aluminum Jug |
£59.95
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965310
|
Thermo Scientific
T001014000 |
4 L | Aluminum Jug |
£90.95
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chromplete Acetonitrile meets the analytical requirements of USP and is suitable for use in:
- HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
- GC- FID, GC- ECD, GC-TCD
- Pesticide Analysis by GC or HPLC
- UV Spectrophotometry
- Preparative chromatography
- Supercritical Fluid and Thin Layer chromatography
- Reactions/Analyses requiring ACS compliance
- Extraction/Concentration Analyses
Chemical Identifiers
75-05-8 | |
41.05 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CHEBI:38472 | |
CC#N |
C2H3N | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
6342 | |
acetonitrile |
Specifications
-51°F | |
75-05-8 | |
≤ 5 APHA | |
180°F | |
1.3405 – 1.3425 | |
C2H3N | |
≤ 0.0006 mEq/g | |
≤ 0.01 % | |
WEVYAHXRMPXWCK-UHFFFAOYSA-N | |
acetonitrile | |
6342 | |
41.05 | |
Chromplete | |
Acetonitrile |
<0.1 at 195 nm, <0.05 at 200 nm, <0.02 at 210 nm, <0.01 at 220 nm | |
0.9995 | |
0.775 - 0.78 | |
Liquid | |
≤ 0.8 ppm | |
≤ 0.008 mEq/g | |
1648 | |
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn | |
CC#N | |
41.05 | |
CHEBI:38472 | |
0.9995 | |
Minimum, GC-FID |
Safety and Handling
Causes serious eye irritation, Highly flammable liquid and vapor, Harmful if swallowed, in contact with skin or if inhaled
Serious eye damage/eye irritation (category 2), Flammable liquids (category 2), Acute toxicity (category 4)
missing translation for 'shelfLife' : 1825 days
missing translation for 'einecsNumber' : 200-835-2
missing translation for 'rtecsNumber' : AL7700000
missing translation for 'storageNote' : In sealed containers
Recommended Storage : Room temperature flammable storage
- Acetonitrile Chromplete
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.