Learn More
Tocris Bioscience™ ci-IP3/PM
Caged inositol triphosphate
Brand: Tocris Bioscience™ 6210/10U
Description
ci-IP3/PM is a caged inositol triphosphate. Cell permeable. UV photolysis (λ = 330 nm) in HeLa cells releases i-IP3, promoting Ca2+-release from internal stores.F 127 (Cat. No. 6253) for the solubilization of ci-IP3/PM is also available.
Specifications
Inositol Triphosphate | |
C42H64NO31P3 | |
Store at -20°C | |
10 μg |
1009832-82-9 | |
1171.87g/mol | |
Caged Compounds |