missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cibisatamab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA542186
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Cibisatamab (CEA-TCB) is a T cell bispecific antibody that binds carcinoembryonic antigen (CEA) on cancer cells and CD3 on T cells. Cibisatamab has a single CD3epsilon chain binding site and two CEA binding sites on T cells (it regulates the binding affinity to cancer cells with high expression of CEA on the cell surface). This avoids targeting healthy epithelial cells with low CEA expression levels. T cell activation, cytokine secretion and release of cytotoxic particles are triggered by binding to CEA on the surface of cancer cells and CD3 on T cells.
Specifications
Cibisatamab | |
Recombinant Monoclonal | |
Unconjugated | |
CEA TCB; RG-7802; RO-6958688 | |
Human CD3E/CEACAM5 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
2.54 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG1 κ, IgG1 λ |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cibisatamab Recombinant Human Monoclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction