Learn More
Tocris Bioscience™ CID 1375606
GPR27 agonist
Brand: Tocris Bioscience™ 6544/5
57.88 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
CID 1375606 is a GPR27 agonist. Promotes recruitment of β-arrestin 2 to chimeric GPR27V2 and wild-type GPR27 in a firefly luciferase complementation assay (pEC50 values are 6.34 and 5.1, respectively).Specifications
2,4-Dichloro-N-(phenylcarbamoyl)phenylbenzamide | |
5 mg | |
98% |
313493-80-0 | |
C20H14Cl2N2O2 |