missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CID 1375606

GPR27 agonist

£72.00 - £274.00

Specifications

Chemical Name or Material 2,4-Dichloro-N-(phenylcarbamoyl)phenylbenzamide
CAS 313493-80-0
Molecular Formula C20H14Cl2N2O2
Purity 98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18795718
View Documents
Tocris Bioscience™
6544/5
5 mg
£72.00
5mg
Please to purchase this item. Need a web account? Register with us today!
18750878
View Documents Promotion Details
Tocris Bioscience™
6544/25
25 mg
£274.00
25mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GPR27 agonist. Promotes recruitment of β-arrestin 2 to chimeric GPR27V2 and wild-type GPR27 in a firefly luciferase complementation assay (pEC50 values are 6.34 and 5.1, respectively).
Specifications

Specifications

2,4-Dichloro-N-(phenylcarbamoyl)phenylbenzamide
313493-80-0
C20H14Cl2N2O2
98%
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications