Learn More
Cilomilast, MedChemExpress
MedChemExpress Cilomilast (SB-207499) is a potent, selective and orally active inhibitor of Phosphodiesterase 4 (PDE4), with IC50s of ∼100 and 120 nM for LPDE4 and HPDE4, respectively.
Brand: Medchem Express HY-10790
This item is not returnable.
View return policy
Description
Cilomilast shows selectivity for PDE4 over PDE1, PDE2, PDE3 and PDE5 (IC50=74, 65, >100, and 83 μM, respectively). Cilomilast has anti-inflammatory and immunomodulatory effects and can be used for thr research of asthma and chronic obstructive pulmonary disease (COPD).Specifications
Cilomilast | |
White | |
C20H25NO4 | |
SB-207499 | |
O=C([C@H]1CC[C@@](C#N)(CC1)C2=CC=C(OC)C(OC3CCCC3)=C2)O | |
343.42 | |
Research | |
COVID-19-immunoregulation |
153259-65-5 | |
Research | |
100 mg | |
DMSO : ≥ 100 mg/mL (291.19 mM) | |
343.42 | |
99.89% | |
Solid |