missing translation for 'onlineSavingsMsg'
Learn More

Cilomilast, MedChemExpress

MedChemExpress Cilomilast (SB-207499) is a potent, selective and orally active inhibitor of Phosphodiesterase 4 (PDE4), with IC50s of ∼100 and 120 nM for LPDE4 and HPDE4, respectively.

£121.00 - £1223.31

Specifications

Chemical Name or Material Cilomilast
CAS 153259-65-5
Color White
Purity Grade Notes Research
Molecular Formula C20H25NO4
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16470507
Encompass_Preferred
View Documents
Medchem Express
HY-10790
50 mg
£694.54
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16464158
Encompass_Preferred
View Documents
Medchem Express
HY-10790
10 mg
£211.75
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16436867
Encompass_Preferred
View Documents
Medchem Express
HY-10790
5 mg
£121.00
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16435126
Encompass_Preferred
View Documents
Medchem Express
HY-10790
100 mg
£1223.31
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cilomilast shows selectivity for PDE4 over PDE1, PDE2, PDE3 and PDE5 (IC50=74, 65, >100, and 83 μM, respectively). Cilomilast has anti-inflammatory and immunomodulatory effects and can be used for thr research of asthma and chronic obstructive pulmonary disease (COPD).
Specifications

Specifications

Cilomilast
White
C20H25NO4
DMSO : ≥ 100 mg/mL (291.19 mM)
343.42
99.89%
Solid
153259-65-5
Research
SB-207499
O=C([C@H]1CC[C@@](C#N)(CC1)C2=CC=C(OC)C(OC3CCCC3)=C2)O
343.42
Research
COVID-19-immunoregulation
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.