missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ cis MZ 1
Negative Control for MZ1
Brand: Tocris Bioscience™ 6155/5
This item is not returnable.
View return policy
Description
Negative Control for MZ 1 (Cat. No. 6154)
cis MZ 1 is a negative Control for MZ1 (Cat.No. 6154). Exhibits no significant VHL binding affinity.Specifications
PROTAC Controls | |
1797406-72-4 | |
1002.64g/mol | |
Bromodomain Inhibitors |
Bromodomain | |
C49H60ClN9O8S2 | |
Store at -20°C | |
5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction