missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ cis VH 298
Negative control for VH 298
Brand: Tocris Bioscience™ 6157/10
This item is not returnable.
View return policy
Description
cis VH 298 is a negative control for VH 298 (Cat.No. 6156). Exhibits no detectable binding of VHL.Specifications
PROTAC Controls | |
C27H33N5O4S | |
Store at -20°C | |
10 mg |
HIF | |
523.65g/mol | |
HIF Activators |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction