missing translation for 'onlineSavingsMsg'
Learn More
Citric acid, trisodium salt, 98%, pure, anhydrous Product Code.: 10362234

Citric acid, trisodium salt, 98%, pure, anhydrous

Product Code. 10362234 Shop All Thermo Scientific Chemicals Products
100 g, Plastic bottle, 100g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10362234

Brand: Thermo Scientific 391971000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68-04-2 | C6H5Na3O7 | 258.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-04-2
Molecular Formula C6H5Na3O7
Molecular Weight (g/mol) 258.07
MDL Number MFCD00012462
InChI Key HRXKRNGNAMMEHJ-UHFFFAOYSA-K
Synonym sodium citrate, trisodium citrate, citrosodine, sodium citrate anhydrous, citrosodina, natrocitral, citnatin, citreme, citrosodna, trisodium citrate, anhydrousShow MoreShow Less
PubChem CID 6224
ChEBI CHEBI:53258
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Melting Point 300.0°C
Color White
Flash Point 290°C
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Plastic bottle
Quantity 100 g
Merck Index 15, 8737
Solubility Information Solubility in water: 500g/L (20°C).
Formula Weight 258.07
Percent Purity 0.98
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Citric acid, trisodium salt
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0476429 Certificate TypeCertificate of Analysis Date10/10/2025 Product Code391971000,391970025,391975000
Lot NumberA0471555 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code391971000,391970025,391975000
Lot NumberA0462445 Certificate TypeCertificate of Analysis Date13/05/2024 Product Code391971000,391970025,391975000
Lot NumberA0458235 Certificate TypeCertificate of Analysis Date29/11/2023 Product Code391971000,391970025,391975000
Lot NumberA0454503 Certificate TypeCertificate of Analysis Date25/08/2023 Product Code391971000,391970025,391975000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Citric acid, trisodium salt, 98%, pure, anhydrous >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.