missing translation for 'onlineSavingsMsg'
Learn More
Citric acid, trisodium salt 98%, anhydrous, pure, Thermo Scientific Chemicals Product Code.: 10360334

Citric acid, trisodium salt 98%, anhydrous, pure, Thermo Scientific Chemicals

Product Code. 10360334
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

Product Code. 10360334

Brand: Thermo Scientific Acros 391975000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68-04-2
Molecular Formula C6H5Na3O7
Molecular Weight (g/mol) 258.07
MDL Number MFCD00012462
InChI Key HRXKRNGNAMMEHJ-UHFFFAOYSA-K
Synonym sodium citrate, trisodium citrate, citrosodine, sodium citrate anhydrous, citrosodina, natrocitral, citnatin, citreme, citrosodna, trisodium citrate, anhydrousShow MoreShow Less
PubChem CID 6224
ChEBI CHEBI:53258
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Melting Point 300.0°C
Color White
Flash Point 290°C
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Plastic bottle
Quantity 500 g
Solubility Information Solubility in water: 500g/L (20°C).
Formula Weight 258.07
Percent Purity 0.98
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material Citric acid, trisodium salt
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462445 Certificate TypeCertificate of Analysis Date13/05/2024 Catalog Number391971000,391970025,391975000
Lot NumberA0458235 Certificate TypeCertificate of Analysis Date29/11/2023 Catalog Number391971000,391970025,391975000
Lot NumberA0454503 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number391971000,391970025,391975000
Lot NumberA0452811 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog Number391971000,391970025,391975000
Lot NumberA0452228 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number391971000,391970025,391975000
Product Identifier
  • Citric acid, trisodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Citric acid, trisodium salt 98%, anhydrous, pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.