Learn More
Sartorius Claristep™ Single Tray
The Claristep™ Tray is a component of the Claristep™ Station. The Station and Tray are used for processing of Claristep™ Filter. The tray of the station can be exchanged and can be ordered separately.
Brand: Sartorius
Code : NEW
Additional Details : Weight : 0.15000kg
Includes: Single Tray only
Description
- Multiplex up to 8 samples
- No vacuum source needed
- No power supply needed
- Yellow
Specifications
600 μL | |
Room Temperature | |
Tools and Accessories | |
8-well Strips |
Single Tray Only | |
HPLC (high-performance liquid chromatography) | |
Analytical Sample Preparation, Clarification |