Learn More
Honeywell™ N7700 Class 1 Half-Mask Respirator
High-performance, half-mask respirator with a lightweight, low-profile design.
Brand:  Honeywell™ N65770033
Additional Details : Weight : 4.10000kg
 View more versions of this product
Product Code. 15866845
  • £31.00 / Each
Estimated Shipment: 24-07-2024
to see stock.
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • 100% Silicone hypoallergenic face piece for extra comfort
  • Single unit four-point harness with neck strap is easy to use and its ergonomic design and improved suspension system fits perfectly on top of the head providing a high level of comfort
  • Reduced dead air space means less re-breathed exhaled air
  • Quick to dismantle and clean, with replaceable parts
  • Superior exhalation valve allows more breath to be exhaled so workers feel cooler and more comfortable
  • Variable thickness in sealing area conforms to nose bridge for improved comfort and fit
Specifications
Specifications
EC Category PPE: 3, EC Certificate: 250, EC Attestation, EN 140
Silicone skirt
Large
4-point
Black
Half-mask Respirator
Suitable for many types of applications