missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ N7700 Class 1 Half-Mask Respirator

High-performance, half-mask respirator with a lightweight, low-profile design.

£31.00 - £43.62

Specifications

Certifications/Compliance EC Category PPE: 3, EC Certificate: 250, EC Attestation, EN 140
Color Black
Material Silicone skirt
Product Type Half-mask Respirator
For Use With (Application) Suitable for many types of applications
View More Specs

Products 2
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15856845
View Documents Promotion Details
Honeywell™
N65770032
Medium
£43.62
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Add to basket
 
15866845
View Documents Promotion Details
Honeywell™
N65770033
Large
£31.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
 
Description

Description

  • 100% Silicone hypoallergenic face piece for extra comfort
  • Single unit four-point harness with neck strap is easy to use and its ergonomic design and improved suspension system fits perfectly on top of the head providing a high level of comfort
  • Reduced dead air space means less re-breathed exhaled air
  • Quick to dismantle and clean, with replaceable parts
  • Superior exhalation valve allows more breath to be exhaled so workers feel cooler and more comfortable
  • Variable thickness in sealing area conforms to nose bridge for improved comfort and fit
Specifications

Specifications

EC Category PPE: 3, EC Certificate: 250, EC Attestation, EN 140
Silicone skirt
Suitable for many types of applications
Black
Half-mask Respirator
4-point
Videos
SDS
Documents

Documents

Product Certifications