missing translation for 'onlineSavingsMsg'
Learn More

Erlab Class 2 Safety Cabinet - Double Ventilation with an Integrated Bio-decontamination System

Solis Premium Class II safety cabinets are designed to protect the handling process, the operator and the environment when working with pathogens.

Brand:  Erlab HOT19847

 View more versions of this product

Product Code. 17776634

  • £15650.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • A highly efficient HEPA H14 filter eliminates particles from the ambient air before supplying air to the handling chamber: 99.999% of particles >0.3 μm are filtered.
  • The extraction rate is 38% and the recirculation rate is 62%.
  • Very strong front air barrier.
  • Electric and sliding front window at a 10° angle made from laminated glass, in compliance with the EN12469 standard.
  • The LED network delivers lighting that can be adjusted from soft to very bright.
Specifications

Specifications

Decontamination with H202
Stainless Steel
Biosafety Cabinet
58 dBA
750 W
>410 m3hr.
Protection of the Handler
Voice Control
USB-A, DOP
290 W
EN 12469-2000
1590 mm
Solis Premium 1500
2
600 mm
Touch Screen
1920 mm
105 W
1506 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.