Learn More
Brand™ Blaubrand™ Eterna™ Class AS Bulb Pipets
Manufactured from AR-GLAS™ Soda-lime glass and complies with DIN EN ISO 648 standards. BRAND™ Blaubrand™ Eterna™ Class AS Bulb Pipets are delivered with batch number and one batch certificate per packing unit of the manufacturer. USP Individual certificate or DAkkS calibration certificate available by request.
Brand:  Brand™ 30515
47.45 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11853980
  • £74.25 / Pack of 6
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from AR-GLAS™ Soda-lime glass.
  • DIN EN ISO 648. Calibrated to deliver (TD, Ex).
  • Waiting time 5 seconds.
  • Delivered with batch number and one batch certificate per packing unit of the manufacturer.
  • On demand also available with individual certificate, USP Individual certificate or DAkkS calibration certificate.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
To Deliver
AS
Yes
DE-M Marking
0.03 mL
25 mL
DIN EN ISO 648, DIN 12691
AR-GLAS, Soda Lime Glass
Amber
6 Pack
Blue
Volumetric Bulb Pipet