missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class E2 Box Weight with COFRAC Calibration Certificate

Calibrate weights of lower accuracy classes and are used with class I weighing instruments. Zwiebel™ Class E2 Box Weights are intended for use with class F1 weights.

Brand:  Zwiebel™ PBI23883

 View more versions of this product

Product Code. 11745203

  • £1700.00 / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Class E2 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
    Specifications

    Specifications

    5 kg
    ±8 mg
    For use with Weighing Instruments of Special Accuracy Class I
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    View accessory products
    Special Offers

    Special Offers