Learn More
Zwiebel™ Class E2 Box Weight with COFRAC Calibration Certificate
Calibrate weights of lower accuracy classes and are used with class I weighing instruments. Zwiebel™ Class E2 Box Weights are intended for use with class F1 weights.
£278.00 - £1810.00
Specifications
For Use With (Application) | For use with Weighing Instruments of Special Accuracy Class I |
---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
11710263
|
Zwiebel™
PBI23733 |
1 to 100 g |
£1810.00
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||||
11715193
|
Zwiebel™
PBI23878 |
100 g |
£410.00
Each |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
11725193
|
Zwiebel™
PBI23879 |
200 g |
£451.00
Each |
Estimated Shipment: 13-01-2025
Log in to see stock. |
|||||
11785183
|
Zwiebel™
PBI23875 |
10 g |
£295.00
Each |
In Stock
Log in to see stock. |
|||||
11755183
|
Zwiebel™
PBI23872 |
1 g |
£278.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
11795183
|
Zwiebel™
PBI23876 |
20 g |
£299.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
11765183
|
Zwiebel™
PBI23873 |
2 g |
£278.00
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
11775183
|
Zwiebel™
PBI23874 |
5 g |
£285.00
Each |
Estimated Shipment: 16-01-2025
Log in to see stock. |
|||||
11705193
|
Zwiebel™
PBI23877 |
50 g |
£318.00
Each |
Estimated Shipment: 17-01-2025
Log in to see stock. |
|||||
11745203
|
Zwiebel™
PBI23883 |
5 kg |
£1720.00
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
Description
Specifications
For use with Weighing Instruments of Special Accuracy Class I |