missing translation for 'onlineSavingsMsg'
Learn More

Zwiebel™ Class E2 Box Weight with COFRAC Calibration Certificate

Calibrate weights of lower accuracy classes and are used with class I weighing instruments. Zwiebel™ Class E2 Box Weights are intended for use with class F1 weights.

Brand:  Zwiebel™ PBI23872

 View more versions of this product

Product Code. 11755183

  • £275.00 / Each
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Class E2 Box Weight
  • Includes COFRAC Calibration Certificate
Specifications

Specifications

1 g
±0.03 mg
For use with Weighing Instruments of Special Accuracy Class I
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers