Learn More
Abeba™ Classic 1011 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
Brand: Abeba™ 1011/36
Additional Details : Weight : 0.88600kg
Description
- CE EN ISO 20345:2011 SB A, E, FO SRC
- Microfiber black
- Steel toe cap
- Slip resistance PU outsole sole
- Sanitized treated
- Antistatic
- Heel strap non-foldable, adjustable
- Microfiber lining and heel patch
- Adjustable instep area
- Buckle fastening
Specifications
CE EN ISO 20345:2011 SB A, E, FO SRC | |
Microfiber | |
36 | |
Polyurethane | |
Steel |
Black | |
Microfiber | |
Slip-resistant | |
1011 | |
Animal fats, oil and fuel |