Learn More
Abeba™ Classic 31000 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
Brand: Abeba™ 31000/45
50.13 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- CE EN ISO 20345:2011 SB SRC
- Grained leather
- White
- Steel toe cap
- Slip resistance PU outsole sole
- ESD dissipative according to DIN EN 61340
- Heel strap non-foldable, adjustable
- Lining microfiber, respiration absorbing
- Adjustable instep area
- Buckle fastening
- Heel patch microfiber
Specifications
CE EN ISO 20345:2011 SB SRC | |
Microfiber | |
45 | |
Polyurethane | |
Steel |
White | |
Leather | |
Slip-resistant | |
31000 | |
Animal fats, oil and fuel |