Learn More
Abeba™ Classic 31000 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
£74.45 - £76.70
Specifications
Certifications/Compliance CE EN ISO 20345:2011 SB SRC
Color White
Liner Material Microfiber
Material Leather
Sole Features Slip-resistant
View More Specs
Products 11
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15465684
View Documents Promotion Details
Abeba™
31000/36
36
£74.45
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
15485684
View Documents Promotion Details
Abeba™
31000/37
37
£74.45
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
15495684
View Documents Promotion Details
Abeba™
31000/38
38
£74.45
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
15405694
View Documents Promotion Details
Abeba™
31000/39
39
£74.45
1 pair
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
15485694
View Documents Promotion Details
Abeba™
31000/40
40
£76.70
1 pair
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
15425704
View Documents Promotion Details
Abeba™
31000/41
41
£76.70
1 pair
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
15445704
View Documents Promotion Details
Abeba™
31000/42
42
£76.70
1 pair
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
15455704
View Documents Promotion Details
Abeba™
31000/43
43
£74.45
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
15475704
View Documents Promotion Details
Abeba™
31000/44
44
£74.45
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
15485704
View Documents Promotion Details
Abeba™
31000/45
45
£74.45
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
15405714
View Documents Promotion Details
Abeba™
31000/46
46
£76.70
1 pair
Estimated Shipment: 02-12-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 SB SRC
  • Grained leather
  • White
  • Steel toe cap
  • Slip resistance PU outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Lining microfiber, respiration absorbing
  • Adjustable instep area
  • Buckle fastening
  • Heel patch microfiber
Specifications
Specifications
CE EN ISO 20345:2011 SB SRC
Microfiber
Slip-resistant
31000
Animal fats, oil and fuel
White
Leather
Polyurethane
Steel