Learn More
ABEBA™ Classic Microfiber Safety Clog OB
ABEBA™ Classic Microfiber Safety Clog OB is made of washable microfiber and a slip resistant PU outsole to provide all day comfort and safety.
Brand:  ABEBA™ 31011/38
Additional Details : Weight : 0.95600kg
 View more versions of this product
Product Code. 15669606
  • £75.87 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • Comfort and safety all day long
  • Comply to EN ISO 20345 : 2012, SB, SRC
  • ESD dissipative according to DIN EN 61340
  • Slip resistant PU outsole
  • Washable black microfiber
  • Adjustable strap for better fitting
  • Steel toe cap (200 joules)
Specifications
Specifications
Stainless steel
Buckles
Microfiber
Microfiber
38
Polyurethane
Steel
EN ISO 20345 : 2012
CE EN ISO 20345:2012 SB SRC ESD
Black
Microfiber
2
Slip-resistant SRC, cushioning, energy absorption
Clog
1 Pair