Learn More
PURUS™ Cleantek CPE Overshoes
Disposable shoe covers provide barrier protection against light spills and dust
Brand:  PURUS™ 48020814
 View more versions of this product
Product Code. 15608643
Description
Description
  • 100 pieces per bag, 20 per case
  • Made from cast polyethelyne
  • helps reduce particulates from entering a cleanroom environment
  • provides protection for shoes against light splash and dust
Specifications
Specifications
Blue
355.6 mm
2,000 Pc./Case
14 in.
Single-use
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
PURUS™ Cleantek CPE Overshoes > Length: 355.6 mm