Learn More
Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates
Perform quantitative and qualitative solid phase immunoassays with these nonsterile 96-well plates, available in a variety of binding surfaces.
£122.00 - £296.00
Specifications
Barcode | No Barcode |
---|---|
Format | Standard |
Lid | Without Lid |
Detection Method | Colorimetric |
Array | 8 x 12 |
Alert:
Each lot is functionally tested for IgG binding.
Product Code | Brand | Surface Treatment | Certified | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Surface Treatment | Certified | Price | Quantity & Availability | ||||
10098860
|
Thermo Scientific™
430341 |
MaxiSorp | No |
£122.00
Pack of 60 |
In Stock
Log in to see stock. |
||||
10737931
|
Thermo Scientific™
446140 |
PolySorp | No |
£296.00
Pack of 60 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
10777621
|
Thermo Scientific™
446612 |
MaxiSorp | Yes |
£184.73
Pack of 60 |
Estimated Shipment: 23-07-2024
Log in to see stock. |
||||
Description
Perform quantitative and qualitative solid phase immunoassays with Thermo Scientific™ Clear C-Shaped Immuno Nonsterile 96-Well Plates, which include Nunc™, Immulon™ and Microtiter™ plates. Choose from a variety of binding surfaces for reliable and reproducible results in colormetric assays.
96-Well Solid Plates:
- Clear polystyrene
- Choice of multiple surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces:
- Thermo Scientific™ PolySorp™ for adsorption of hydrophobic molecules
- Thermo Scientific™ MaxiSorp™ is hydrophilic and ideal for antibody sandwich assays
Ordering Information:
Please note that availability may vary by country. 446612 includes certificate.
Specifications
No Barcode | |
Without Lid | |
8 x 12 | |
Clear | |
Polystyrene | |
250 μL | |
Round, C-bottom | |
5 | |
C96 |
Standard | |
Colorimetric | |
60 Cases | |
Microplate | |
96 | |
Non-sterile | |
60 | |
None |