Learn More
Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates
GREENER_CHOICE
Choose from four binding surfaces of these 96-well plates for reliable and reproducible results in colormetric assays.
Brand:  Thermo Scientific™ 3355
91.48 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
 View more versions of this product
Product Code. 10547915
  • £151.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Alert:
Each lot is functionally tested for IgG binding.
Description
Description
Choose from four binding surfaces for reliable and reproducible results in colormetric assays with Thermo Scientific™ Clear Flat-Bottom Immuno Nonsterile 96-Well Plates, which feature Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates:
  • Clear polystyrene
  • Choice of four surfaces for optimal binding
  • Alphanumerically labeled rows and columns
Surfaces:
  • Thermo Scientific PolySorp, Immulon 1B, and Universal Binding (UB) are for adsorption of hydrophobic molecules.
  • Thermo Scientific Medisorp and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules.
  • Thermo Scientific MaxiSorp, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays.
  • Thermo Scientific MultiSorp is very hydrophilic and works well for glycans and water soluble proteins.
Ordering Information:
Please note that availability may vary by country. 467320, 456537, 460984 and 439454 include certificate
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Barcode
Standard
8 x 12
Hydrophobic
Microplate
330 μL
230 μL
Immulon 1 B
50
None
No
Colorimetric
50 Cases
Clear
Polystyrene
96
Non-sterile
Round Bottom (U-well)
10
Lipids