Learn More
Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates
Carry out colormetric assays with reliable and reproducible results using these clear, round-bottom, immuno nonsterile 96-well plates, offered with a choice of surface.
Brand: Thermo Scientific™ 3555
106.87 GBP valid until 2024-12-31
Use promo code "23291" to get your promotional price.
Code : 04
Additional Details : Weight : 2.43000kg
Alert:
Each lot is functionally tested for IgG binding.
Description
Carry out colormetric assays with reliable and reproducible results using Thermo Scientific™ Clear Round-Bottom Immuno Nonsterile 96-Well Plates, offered with a choice of surface for optimal binding. Features Thermo Scientific™ Nunc™, Immulon™ and Microtiter™ clear plates that are ideal for quantitative and qualitative solid phase immunoassays.
96-Well Solid Plates
- Clear polystyrene
- Choice of four surfaces for optimal binding
- Alphanumerically labeled rows and columns
Surfaces
- PolySorp and Immulon 1B for adsorption of hydrophobic molecules
- Immulon 2HB is slightly hydrophilic and binds a diverse range of biomolecules
- MaxiSorp is hydrophilic and ideal for antibody sandwich assays
Ordering Information:
Please note that availability may vary by country.
Specifications
No Barcode | |
Standard | |
8 x 12 | |
Clear | |
Polystyrene | |
96 | |
Non-sterile | |
Round | |
10 | |
Lipids |
Hydrophobic | |
Colorimetric | |
50 Cases | |
Microplate | |
280 μL | |
180 μL | |
Immulon 1 B | |
50 | |
None |