missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diversey™ ClearKlens Cleansinald Sterile VH9S Disinfectant
Ready to use disinfectant with WFI
Brand: Diversey™ 100848254
Additional Details : Weight : 5.40000kg
Description
ClearKlens Cleansinald is a sterile ready to use surface disinfectant for pharmaceutical cleanrooms.
Key properties
- ClearKlens Cleansinald is a ready to use blend of quaternary ammonium and alkyl amines diluted to 0.5% v/v with Water for Injection (WFI).
- ClearKlens Cleansinald is used for cleaning and disinfection of walls, floors, work surfaces, and process equipment in pharmaceutical industry.
- ClearKlens Cleansinald concentrate is diluted at 0.5% with water for injection (WFI), according to the European Pharmacopeia, and then filtered at 0.22 μm.
Benefits
- Ready to use
- Broad spectrum of activity
- Low toxicity
- 0.22 μm filtered
- Delivered with conformity certificates
Specifications
9 to 11.4 | |
Ready to use blend of quaternary ammonium and alkyl amines diluted to 0.5% v/v with Water for Injection | |
900 mL |
Sterile | |
6/Cs. |