Learn More
Nickel Electro™ Clifton™ Bath Lab Armor™
Obtain superior water bath cleaning results when you replace water in your water bath with these thermally efficient pellets that are easy to keep clean. This Clifton™ Bath Armor™ tackles water contamination and protects your water bath from drying out.
Brand: Nickel Electro™
Code : 03
Additional Details : Weight : 1.00000kg
Description
- Tackles water contamination by utilizing a dry, metallic thermal media comprised of small beads instead of water
- Thermally efficient pellets that are easy to keep clean
- Moisture and gas impermeable
- High thermal conductivity
- Smooth and rounded almost fluid like
- Reduces water consumption
- Saves the need for racks, floaties and bottle weights
- Do not autoclave
Specifications
0.5 L | |
Bath Lab Armor | |
0.5 L |